Product Details

SNP ID
rs149982395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:1090137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCCTTGGTCATGACGACGTGCTC[A/G]GGGACTAGCTGCCGAGAGAGGAAGC
Phenotype
MIM: 601155 MIM: 180664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGAP45 PubMed Links

Gene Details

Gene
ARHGAP45
Gene Name
Rho GTPase activating protein 45
There are no transcripts associated with this gene.

Gene
POLR2E
Gene Name
RNA polymerase II subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001316323.1 329 Silent Mutation CCC,CCT P53P NP_001303252.1
NM_001316324.1 329 Silent Mutation CCC,CCT P53P NP_001303253.1
NM_002695.3 329 Silent Mutation CCC,CCT P146P NP_002686.2
XM_011528070.2 329 Silent Mutation CCC,CCT P146P XP_011526372.1
XM_017026883.1 329 Silent Mutation CCC,CCT P53P XP_016882372.1

View Full Product Details