Product Details

SNP ID
rs150281427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3186168 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCCGCCGACGCCGCGCACGAGTT[C/T]ACCGTGTACCGCATGCAGCAGTACG
Phenotype
MIM: 609156 MIM: 603751
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NCLN PubMed Links

Gene Details

Gene
NCLN
Gene Name
nicalin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321463.1 292 Silent Mutation TTC,TTT F46F NP_001308392.1
NM_020170.3 292 Silent Mutation TTC,TTT F46F NP_064555.2
Gene
S1PR4
Gene Name
sphingosine-1-phosphate receptor 4
There are no transcripts associated with this gene.

View Full Product Details