Product Details

SNP ID
rs150349860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:32598200 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGGAGAGGGGGCCAGCAGCCTCC[C/G]GGCCCTGGGACACGACCGCATCCTC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANKRD27 PubMed Links

Gene Details

Gene
ANKRD27
Gene Name
ankyrin repeat domain 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032139.2 3254 Missense Mutation CCG,CGG P1033R NP_115515.2
XM_006723412.2 3254 Missense Mutation CCG,CGG P1033R XP_006723475.1
XM_017027349.1 3254 Missense Mutation CCG,CGG P837R XP_016882838.1

View Full Product Details