Product Details

SNP ID
rs151181290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:2076962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACGTGCACGAACACGCGGAACAGC[C/T]GCGACAGGATCTTCCGCACCGTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MOB3A PubMed Links

Gene Details

Gene
MOB3A
Gene Name
MOB kinase activator 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130807.2 791 Missense Mutation CAG,CGG Q158R NP_570719.1
XM_011527683.2 791 Missense Mutation CAG,CGG Q158R XP_011525985.1

View Full Product Details