Product Details

SNP ID
rs111480471
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:29241085 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGGACACATGTGACCTGTTAGG[A/G]TGTCTAGCCATCTGGTTATCGTGTG
Phenotype
MIM: 602454
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPRU PubMed Links

Gene Details

Gene
PTPRU
Gene Name
protein tyrosine phosphatase, receptor type U
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195001.1 Intron NP_001181930.1
NM_005704.4 Intron NP_005695.3
NM_133177.3 Intron NP_573438.3
NM_133178.3 Intron NP_573439.2
XM_006710269.2 Intron XP_006710332.1
XM_016999992.1 Intron XP_016855481.1
XM_016999993.1 Intron XP_016855482.1
XM_016999994.1 Intron XP_016855483.1
XM_016999995.1 Intron XP_016855484.1

View Full Product Details