Product Details

SNP ID
rs111953674
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:23692607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGACTGCTGCTCTTCCCTGTTGC[A/C]GCAGGATCAAGGTGAAAAGGAGAAC
Phenotype
MIM: 604175
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPL11 PubMed Links

Gene Details

Gene
RPL11
Gene Name
ribosomal protein L11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000975.3 Intron NP_000966.2
NM_001199802.1 Intron NP_001186731.1

View Full Product Details