Product Details

SNP ID
rs112004526
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:206866334 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGAAACATTGACATCAGAATCTT[A/G]AGGAGGACTGAGTCTTTGCAAGACA
Phenotype
MIM: 605619
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IL20 PubMed Links

Gene Details

Gene
IL20
Gene Name
interleukin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018724.3 665 Silent Mutation TTA,TTG L65L NP_061194.2
XM_005273152.3 665 Missense Mutation TTA,TTG L150L XP_005273209.1

View Full Product Details