Product Details

SNP ID
rs114233412
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:45327962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTAACATTGTGGAGGCCACTGAG[A/G]GGGTGGCAACTGCTGGAGGCCAGTT
Phenotype
MIM: 604933
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HPDL PubMed Links

Gene Details

Gene
HPDL
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032756.2 1090 Missense Mutation AGG,GGG R272G NP_116145.1
Gene
MUTYH
Gene Name
mutY DNA glycosylase
There are no transcripts associated with this gene.

View Full Product Details