Product Details

SNP ID
rs114328241
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:3752717 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGCATGGCCAGCCGGGACGCCC[A/G]GTGCCCAGAATGGAAACCGCACCAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC27 PubMed Links

Gene Details

Gene
CCDC27
Gene Name
coiled-coil domain containing 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152492.2 320 Missense Mutation CAG,CGG Q79R NP_689705.2
XM_011540780.1 320 Missense Mutation CAG,CGG Q79R XP_011539082.1
XM_017000402.1 320 Missense Mutation CAG,CGG Q79R XP_016855891.1
XM_017000403.1 320 Missense Mutation CAG,CGG Q79R XP_016855892.1
XM_017000404.1 320 Missense Mutation CAG,CGG Q79R XP_016855893.1
XM_017000405.1 320 Missense Mutation CAG,CGG Q79R XP_016855894.1
XM_017000406.1 320 UTR 5 XP_016855895.1
Gene
TP73-AS1
Gene Name
TP73 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details