Product Details

SNP ID
rs114659399
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:228511679 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACCTGCACCTGCACAGTGGCCTC[A/G]CCGAACTTTACACCATCCTTGAAGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BTNL10 PubMed Links

Gene Details

Gene
BTNL10
Gene Name
butyrophilin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287262.1 432 Silent Mutation NP_001274191.1
XM_011544015.2 432 Silent Mutation XP_011542317.1
XM_016999986.1 432 Silent Mutation XP_016855475.1
XM_016999987.1 432 Silent Mutation XP_016855476.1

View Full Product Details