Product Details

SNP ID
rs115984632
Assay Type
Functionally tested
NCBI dbSNP Submissions
8
Location
Chr.1:205768497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCATCCCTAGAAATTCTGATTC[A/C]GTAGGTTTGAGGTCGGGACCCAAAA
Phenotype
MIM: 603949
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RAB29 PubMed Links

Gene Details

Gene
RAB29
Gene Name
RAB29, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135662.1 2403 UTR 3 NP_001129134.1
NM_001135663.1 2403 UTR 3 NP_001129135.1
NM_001135664.1 2403 UTR 3 NP_001129136.1
NM_003929.2 2403 UTR 3 NP_003920.1
XM_005245569.1 2403 UTR 3 XP_005245626.1
XM_005245570.1 2403 UTR 3 XP_005245627.1
XM_005245571.1 2403 UTR 3 XP_005245628.1
XM_006711605.3 2403 UTR 3 XP_006711668.1
XM_006711606.2 2403 UTR 3 XP_006711669.1
XM_017002748.1 2403 UTR 3 XP_016858237.1
XM_017002749.1 2403 UTR 3 XP_016858238.1
XM_017002750.1 2403 UTR 3 XP_016858239.1

View Full Product Details