Product Details

SNP ID
rs116054135
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:40489177 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCACCAGAATCTATACCGAGA[G/T]GTGATGCTGGAGAACTACAGCAACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ZFP69 PubMed Links

Gene Details

Gene
ZFP69
Gene Name
ZFP69 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320178.1 1310 Missense Mutation GAG,GAT E103D NP_001307107.1
NM_001320179.1 1310 Missense Mutation GAG,GAT E103D NP_001307108.1
NM_198494.2 1310 Missense Mutation GAG,GAT E103D NP_940896.2
XM_005270809.4 1310 Missense Mutation GAG,GAT E103D XP_005270866.1
XM_006710605.3 1310 Missense Mutation GAG,GAT E103D XP_006710668.1
XM_006710606.3 1310 Missense Mutation GAG,GAT E103D XP_006710669.1

View Full Product Details