Product Details

SNP ID
rs116419914
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:40489166 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGGCTCCTGCTCACCAGAAT[C/G]TATACCGAGAGGTGATGCTGGAGAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZFP69 PubMed Links

Gene Details

Gene
ZFP69
Gene Name
ZFP69 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320178.1 1299 Missense Mutation CTA,GTA L100V NP_001307107.1
NM_001320179.1 1299 Missense Mutation CTA,GTA L100V NP_001307108.1
NM_198494.2 1299 Missense Mutation CTA,GTA L100V NP_940896.2
XM_005270809.4 1299 Missense Mutation CTA,GTA L100V XP_005270866.1
XM_006710605.3 1299 Missense Mutation CTA,GTA L100V XP_006710668.1
XM_006710606.3 1299 Missense Mutation CTA,GTA L100V XP_006710669.1

View Full Product Details