Product Details

SNP ID
rs116571455
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:53197277 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCACAATCTCCTAGCACAGGAAC[C/T]TCAGAACCCTCAAATTCTCTTCCAG
Phenotype
MIM: 600650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPT2 PubMed Links

Gene Details

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 Intron NP_000089.1
XM_005270484.1 Intron XP_005270541.1

View Full Product Details