Product Details

SNP ID
rs117316015
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:178548622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAAACTGCAGGTCCCTGTAATGA[C/T]TTCAAAGACTCCGTTCCAACAATGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf220 PubMed Links

Gene Details

Gene
C1orf220
Gene Name
chromosome 1 open reading frame 220
There are no transcripts associated with this gene.

View Full Product Details