Product Details

SNP ID
rs137986343
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:1331478 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGGGGGGGCTGTTCCCCCTGGGC[A/G]AGGCCGAGGAGGCTGGCCTCCGCAG
Phenotype
MIM: 611354 MIM: 615467 MIM: 601365 MIM: 605865
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPSF3L PubMed Links

Gene Details

Gene
CPSF3L
Gene Name
cleavage and polyadenylation specific factor 3-like
There are no transcripts associated with this gene.

Gene
CPTP
Gene Name
ceramide-1-phosphate transfer protein
There are no transcripts associated with this gene.

Gene
DVL1
Gene Name
dishevelled segment polarity protein 1
There are no transcripts associated with this gene.

Gene
MIR6808
Gene Name
microRNA 6808
There are no transcripts associated with this gene.

Gene
TAS1R3
Gene Name
taste 1 receptor member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152228.2 133 Missense Mutation AAG,GAG K45E NP_689414.1
XM_017002435.1 133 Missense Mutation AAG,GAG K45E XP_016857924.1
XM_017002436.1 133 Missense Mutation AAG,GAG K45E XP_016857925.1

View Full Product Details