Product Details

SNP ID
rs138235832
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:153661268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGCAGGAGAGCAATGCTGGATTC[A/G]GGAATTTACCCCCCTGGCTCCCCAG
Phenotype
MIM: 603181 MIM: 607007
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILF2 PubMed Links

Gene Details

Gene
ILF2
Gene Name
interleukin enhancer binding factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267809.1 468 Intron NP_001254738.1
NM_004515.3 468 Intron NP_004506.2
Gene
SNAPIN
Gene Name
SNAP associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012437.5 468 Silent Mutation TCA,TCG S126S NP_036569.1

View Full Product Details