Product Details

SNP ID
rs138749967
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:84479314 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCCGGGGATAAGAGCAGCAGCAG[C/T]GGGAAGAAAAGTCTAAAACGGAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPF1 PubMed Links

Gene Details

Gene
RPF1
Gene Name
ribosome production factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025065.6 78 Silent Mutation AGC,AGT S11S NP_079341.2

View Full Product Details