Product Details

SNP ID
rs139354227
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:162645690 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGTTGAACAAATTGTTATTAATA[G/T]CTTTGAGGGTTCATAAGTTCAGGAA
Phenotype
MIM: 191311
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DDR2 PubMed Links

Gene Details

Gene
DDR2
Gene Name
discoidin domain receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014796.1 Intron NP_001014796.1
NM_006182.2 Intron NP_006173.2
XM_006711344.3 Intron XP_006711407.1
XM_011509586.2 Intron XP_011507888.1
XM_011509587.2 Intron XP_011507889.1
XM_011509588.2 Intron XP_011507890.1

View Full Product Details