Product Details

SNP ID
rs140194675
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:53200790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACAGAAGCCTCTCTTGAATGATG[A/G]CCAGTTCAGGTAAACACTGAGAACC
Phenotype
MIM: 600650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPT2 PubMed Links

Gene Details

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 407 Missense Mutation GAC,GGC D75G NP_000089.1
XM_005270484.1 407 Missense Mutation GAC,GGC D75G XP_005270541.1

View Full Product Details