Product Details

SNP ID
rs140740104
Assay Type
Functionally Tested
NCBI dbSNP Submissions
8
Location
Chr.1:58576436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCGCGCACGCGCAAGTCCAGGC[C/T]GCCGCGGCCCTGGAATAGAGACTCG
Phenotype
MIM: 137290
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TACSTD2 PubMed Links

Gene Details

Gene
TACSTD2
Gene Name
tumor-associated calcium signal transducer 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002353.2 1059 Missense Mutation AGC,GGC S241G NP_002344.2

View Full Product Details