Product Details

SNP ID
rs140866709
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:30732853 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCTCGGGCAGAACAGATTCGCCT[A/G]TGACTGAATCGACTGAATGGATGGC
Phenotype
MIM: 601476 MIM: 115437
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LAPTM5 PubMed Links

Gene Details

Gene
LAPTM5
Gene Name
lysosomal protein transmembrane 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006762.2 1688 UTR 3 NP_006753.1
XM_011542098.2 1688 UTR 3 XP_011540400.1
Gene
MATN1
Gene Name
matrilin 1, cartilage matrix protein
There are no transcripts associated with this gene.

Gene
MATN1-AS1
Gene Name
MATN1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
MIR4420
Gene Name
microRNA 4420
There are no transcripts associated with this gene.

View Full Product Details