Product Details

SNP ID
rs141031190
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:210238062 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACACTATGGGAGGCTGACAGCTA[C/T]GGAGGCCCAAGCCCCCCAGGGCCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SERTAD4 PubMed Links

Gene Details

Gene
SERTAD4
Gene Name
SERTA domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019605.3 481 Silent Mutation TAC,TAT Y34Y NP_062551.1
XM_005273193.4 481 Silent Mutation TAC,TAT Y34Y XP_005273250.1
Gene
SERTAD4-AS1
Gene Name
SERTAD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details