Product Details

SNP ID
rs142236639
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:151518558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAACATGGCTGAGCCCCGGGGCCC[C/T]GTAGACCATGGAGTCCAGATTCGCT
Phenotype
MIM: 609473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CGN PubMed Links

Gene Details

Gene
CGN
Gene Name
cingulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020770.2 183 Silent Mutation CCC,CCT P13P NP_065821.1
XM_005245365.4 183 Silent Mutation CCC,CCT P13P XP_005245422.1

View Full Product Details