Product Details

SNP ID
rs142487972
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:206866308 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCTTCTGTTTAGCAAGCCAAA[G/T]ATGGAAACATTGACATCAGAATCTT
Phenotype
MIM: 605619
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IL20 PubMed Links

Gene Details

Gene
IL20
Gene Name
interleukin 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018724.3 639 Missense Mutation GAT,TAT D57Y NP_061194.2
XM_005273152.3 639 Missense Mutation GAT,TAT D142Y XP_005273209.1

View Full Product Details