Product Details

SNP ID
rs142598568
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:53200761 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAAGACACCATTAGGAGATACCT[C/G]AGTGCACAGAAGCCTCTCTTGAATG
Phenotype
MIM: 600650
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CPT2 PubMed Links

Gene Details

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 378 Silent Mutation CTC,CTG L65L NP_000089.1
XM_005270484.1 378 Silent Mutation CTC,CTG L65L XP_005270541.1

View Full Product Details