Product Details

SNP ID
rs143116225
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:210237972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGCTGTCAGAGATGACTCTGGT[A/T]CTGTCCATGAATAGATTCTGCGAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
SERTAD4 PubMed Links

Gene Details

Gene
SERTAD4
Gene Name
SERTA domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019605.3 391 Silent Mutation GTA,GTT V4V NP_062551.1
XM_005273193.4 391 Silent Mutation GTA,GTT V4V XP_005273250.1
Gene
SERTAD4-AS1
Gene Name
SERTAD4 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details