Product Details

SNP ID
rs143207556
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:94532378 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCGGGCTGTCTGTACTCTTCC[A/G]GTTAACTGTTCGGGAGGGAATCACT
Phenotype
MIM: 134390
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
F3 PubMed Links

Gene Details

Gene
F3
Gene Name
coagulation factor III, tissue factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178096.1 915 Intron NP_001171567.1
NM_001993.4 915 Missense Mutation NP_001984.1

View Full Product Details