Product Details

SNP ID
rs143482127
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:119420181 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTGTCCAGAATACAATCTCTTC[A/G]GCTCACCACAGGGTCTACTATTACA
Phenotype
MIM: 613890
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSD3B2 PubMed Links

Gene Details

Gene
HSD3B2
Gene Name
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000198.3 Intron NP_000189.1
NM_001166120.1 Intron NP_001159592.1

View Full Product Details