Product Details

SNP ID
rs143584741
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:46184323 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGGGCTACTTCCTCATCGCAGCC[A/G]GCGTTGTGGTCTTTGCTCTTGGTTT
Phenotype
MIM: 606076 MIM: 606822 MIM: 613170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIK3R3 PubMed Links

Gene Details

Gene
PIK3R3
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
There are no transcripts associated with this gene.

Gene
POMGNT1
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
There are no transcripts associated with this gene.

Gene
TSPAN1
Gene Name
tetraspanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005727.3 638 Missense Mutation AGC,GGC S64G NP_005718.2
XM_011540460.2 638 Missense Mutation AGC,GGC S64G XP_011538762.1

View Full Product Details