Product Details

SNP ID
rs144405495
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:54036666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAACAGTTGCACAACAAATCCAAA[A/G]CAATACCATCACCGAGAGGACAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM59 PubMed Links

Gene Details

Gene
TMEM59
Gene Name
transmembrane protein 59
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305043.1 683 Missense Mutation NP_001291972.1
NM_001305049.1 683 Missense Mutation NP_001291978.1
NM_001305050.1 683 Missense Mutation NP_001291979.1
NM_001305051.1 683 Missense Mutation NP_001291980.1
NM_001305052.1 683 Missense Mutation NP_001291981.1
NM_001305066.1 683 Intron NP_001291995.1
NM_004872.4 683 Missense Mutation NP_004863.2
XM_006711051.1 683 Missense Mutation XP_006711114.1

View Full Product Details