Product Details

SNP ID
rs144798109
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:28150287 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCTGCACCACGTGGTGGGGCAC[A/G]AAGCAGATGATGAACACCGCCAAGA
Phenotype
MIM: 173393
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTAFR PubMed Links

Gene Details

Gene
PTAFR
Gene Name
platelet activating factor receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000952.4 1070 Silent Mutation TTC,TTT F245F NP_000943.1
NM_001164721.1 1070 Silent Mutation TTC,TTT F245F NP_001158193.1
NM_001164722.2 1070 Silent Mutation TTC,TTT F245F NP_001158194.1
NM_001164723.2 1070 Silent Mutation TTC,TTT F245F NP_001158195.1

View Full Product Details