Product Details

SNP ID
rs144865741
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:213051468 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTACACTGTCACCGAGCCCCAG[C/G]GACACCCGAGGGGCTACACAGTATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RPS6KC1 PubMed Links

Gene Details

Gene
RPS6KC1
Gene Name
ribosomal protein S6 kinase C1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136138.2 217 Missense Mutation CGA,GGA R22G NP_001129610.1
NM_001287218.1 217 UTR 5 NP_001274147.1
NM_001287219.1 217 UTR 5 NP_001274148.1
NM_001287220.1 217 UTR 5 NP_001274149.1
NM_001287221.1 217 UTR 5 NP_001274150.1
NM_012424.4 217 Missense Mutation CGA,GGA R22G NP_036556.2
XM_005273095.1 217 Missense Mutation CGA,GGA R22G XP_005273152.1
XM_005273096.1 217 UTR 5 XP_005273153.1
XM_006711272.1 217 Missense Mutation CGA,GGA R22G XP_006711335.1
XM_017001024.1 217 Intron XP_016856513.1
XM_017001025.1 217 UTR 5 XP_016856514.1
XM_017001026.1 217 UTR 5 XP_016856515.1
XM_017001027.1 217 UTR 5 XP_016856516.1
XM_017001028.1 217 UTR 5 XP_016856517.1
XM_017001029.1 217 UTR 5 XP_016856518.1
XM_017001030.1 217 UTR 5 XP_016856519.1
XM_017001031.1 217 UTR 5 XP_016856520.1
XM_017001032.1 217 UTR 5 XP_016856521.1
XM_017001033.1 217 UTR 5 XP_016856522.1
XM_017001034.1 217 UTR 5 XP_016856523.1

View Full Product Details