Product Details

SNP ID
rs145776320
Assay Type
Functionally Tested
NCBI dbSNP Submissions
22
Location
Chr.1:84479292 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGGAGCCAAGACCATGGCGAAAG[C/T]CGGGGATAAGAGCAGCAGCAGCGGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
RPF1 PubMed Links

Gene Details

Gene
RPF1
Gene Name
ribosome production factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025065.6 56 Missense Mutation GCC,GTC A4V NP_079341.2

View Full Product Details