Product Details

SNP ID
rs145926853
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:154569488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGTGGGGTACGGATACAGAGGAG[C/T]GGCTGGTGGAGCATCTCCTGGATCC
Phenotype
MIM: 118507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 337 Missense Mutation CGG,TGG R31W NP_000739.1
XM_017000180.1 337 UTR 5 XP_016855669.1

View Full Product Details