Product Details

SNP ID
rs146159244
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:53200784 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCAGTGCACAGAAGCCTCTCTTGA[A/G]TGATGGCCAGTTCAGGTAAACACTG
Phenotype
MIM: 600650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CPT2 PubMed Links

Gene Details

Gene
CPT2
Gene Name
carnitine palmitoyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000098.2 401 Missense Mutation AAT,AGT N73S NP_000089.1
XM_005270484.1 401 Missense Mutation AAT,AGT N73S XP_005270541.1

View Full Product Details