Product Details
- SNP ID
-
rs146503193
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
4
- Location
-
Chr.1:156816045 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTCTGGCAGCTCCTGCGGGTCAT[A/G]TCTGTGATCTGGAAGGTGCTGAAGG
- Phenotype
-
MIM: 191315
MIM: 604514
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NTRK1
PubMed Links
Gene Details
- Gene
- NTRK1
- Gene Name
- neurotrophic receptor tyrosine kinase 1
- Gene
- SH2D2A
- Gene Name
- SH2 domain containing 2A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001161441.1 |
194 |
Silent Mutation |
GAC,GAT |
D28D |
NP_001154913.1 |
NM_001161442.1 |
194 |
Silent Mutation |
GAC,GAT |
D10D |
NP_001154914.1 |
NM_001161443.1 |
194 |
UTR 5 |
|
|
NP_001154915.1 |
NM_001161444.1 |
194 |
Silent Mutation |
GAC,GAT |
D28D |
NP_001154916.1 |
NM_003975.3 |
194 |
Silent Mutation |
GAC,GAT |
D28D |
NP_003966.2 |
XM_006711615.2 |
194 |
Silent Mutation |
GAC,GAT |
D34D |
XP_006711678.1 |
XM_017002762.1 |
194 |
Silent Mutation |
GAC,GAT |
D34D |
XP_016858251.1 |
XM_017002763.1 |
194 |
Silent Mutation |
GAC,GAT |
D34D |
XP_016858252.1 |
XM_017002764.1 |
194 |
Silent Mutation |
GAC,GAT |
D28D |
XP_016858253.1 |
XM_017002765.1 |
194 |
Silent Mutation |
GAC,GAT |
D10D |
XP_016858254.1 |
XM_017002766.1 |
194 |
Silent Mutation |
GAC,GAT |
D10D |
XP_016858255.1 |
XM_017002767.1 |
194 |
Silent Mutation |
GAC,GAT |
D34D |
XP_016858256.1 |
View Full Product Details