Product Details

SNP ID
rs146876203
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:228510527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGCCCAGAGCCTGGTGGTGGC[C/T]GAGGCTGATAGATTGGTCACAGCAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BTNL10 PubMed Links

Gene Details

Gene
BTNL10
Gene Name
butyrophilin like 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287262.1 612 Silent Mutation TCA,TCG S153S NP_001274191.1
XM_011544015.2 612 Silent Mutation TCA,TCG S153S XP_011542317.1
XM_016999986.1 612 Silent Mutation TCA,TCG S153S XP_016855475.1
XM_016999987.1 612 Silent Mutation TCA,TCG S153S XP_016855476.1

View Full Product Details