Product Details

SNP ID
rs146920112
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:151728802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCCAAGACTTCCCAACAAGATTC[A/G]CATGCAGCTAATTAAAGACAAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RIIAD1 PubMed Links

Gene Details

Gene
RIIAD1
Gene Name
regulatory subunit of type II PKA R-subunit (RIIa) domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144956.1 245 Missense Mutation CAC,CGC H82R NP_001138428.1

View Full Product Details