Product Details

SNP ID
rs147555011
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:26794427 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTTGTTCTTCATGTTGGCTGC[A/G]GTTGCACTTCATGACCTGGGTTGTC
Phenotype
MIM: 610274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIGV PubMed Links

Gene Details

Gene
PIGV
Gene Name
phosphatidylinositol glycan anchor biosynthesis class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202554.1 1082 Silent Mutation GCA,GCG A131A NP_001189483.1
NM_017837.3 1082 Silent Mutation GCA,GCG A131A NP_060307.2

View Full Product Details