Product Details

SNP ID
rs147678668
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:220151678 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCATTCCTTCTTTTGTCTGGGT[G/T]TGGAGAAGCGCATGAGCCAGCCTTT
Phenotype
MIM: 612801 MIM: 609275
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
RAB3GAP2
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012414.3 4118 Missense Mutation CAA,CAC Q1318H NP_036546.2

View Full Product Details