Product Details
- SNP ID
-
rs147744090
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:205770795 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATGTTTCTGTCCAACCTGTGAAACC[A/G]TTCTCTTTACTGAACCGGTCAATCT
- Phenotype
-
MIM: 603949
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RAB29
PubMed Links
Gene Details
- Gene
- RAB29
- Gene Name
- RAB29, member RAS oncogene family
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001135662.1 |
384 |
Silent Mutation |
AAC,AAT |
N146N |
NP_001129134.1 |
NM_001135663.1 |
384 |
Silent Mutation |
AAC,AAT |
N122N |
NP_001129135.1 |
NM_001135664.1 |
384 |
Silent Mutation |
AAC,AAT |
N74N |
NP_001129136.1 |
NM_003929.2 |
384 |
Silent Mutation |
AAC,AAT |
N146N |
NP_003920.1 |
XM_005245569.1 |
384 |
Silent Mutation |
AAC,AAT |
N146N |
XP_005245626.1 |
XM_005245570.1 |
384 |
Silent Mutation |
AAC,AAT |
N146N |
XP_005245627.1 |
XM_005245571.1 |
384 |
Silent Mutation |
AAC,AAT |
N146N |
XP_005245628.1 |
XM_006711605.3 |
384 |
Silent Mutation |
AAC,AAT |
N74N |
XP_006711668.1 |
XM_006711606.2 |
384 |
Silent Mutation |
AAC,AAT |
N74N |
XP_006711669.1 |
XM_017002748.1 |
384 |
Silent Mutation |
AAC,AAT |
N122N |
XP_016858237.1 |
XM_017002749.1 |
384 |
Silent Mutation |
AAC,AAT |
N122N |
XP_016858238.1 |
XM_017002750.1 |
384 |
Silent Mutation |
AAC,AAT |
N122N |
XP_016858239.1 |
View Full Product Details