Product Details

SNP ID
rs147790749
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:25245254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCAAGTCAATGTTGGTTGTTCCTA[A/G]AGCTTTCGCTGCATTGGTTTTTGCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RSRP1 PubMed Links

Gene Details

Gene
RSRP1
Gene Name
arginine and serine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321772.1 810 Silent Mutation CTA,TTA L190L NP_001308701.1
NM_020317.4 810 Silent Mutation CTA,TTA L190L NP_064713.3
XM_011541797.1 810 Silent Mutation CTA,TTA L190L XP_011540099.1

View Full Product Details