Product Details

SNP ID
rs149098565
Assay Type
Functionally Tested
NCBI dbSNP Submissions
2
Location
Chr.1:45327184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCCTTCGTTTGTGCCACATCGC[C/T]TTCCACGTGCCCGCCGGGCAGCCCC
Phenotype
MIM: 604933
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
HPDL PubMed Links

Gene Details

Gene
HPDL
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032756.2 312 Silent Mutation GCC,GCT A12A NP_116145.1
Gene
MUTYH
Gene Name
mutY DNA glycosylase
There are no transcripts associated with this gene.

View Full Product Details