Product Details
- SNP ID
-
rs149133861
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:26361689 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCTGCCCCATCACCAGGGGAGTCC[C/T]GGCACTGCTCAGGCTCACTGCTCTT
- Phenotype
-
MIM: 616775
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AIM1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs11583897] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AIM1L
- Gene Name
- absent in melanoma 1-like
There are no transcripts associated with this gene.
- Gene
- ZNF683
- Gene Name
- zinc finger protein 683
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001114759.2 |
1794 |
Missense Mutation |
AGG,GGG |
R493G |
NP_001108231.1 |
NM_001307925.1 |
1794 |
Missense Mutation |
AGG,GGG |
R513G |
NP_001294854.1 |
NM_173574.3 |
1794 |
Missense Mutation |
AGG,GGG |
R493G |
NP_775845.2 |
XM_005245828.3 |
1794 |
Missense Mutation |
AGG,GGG |
R521G |
XP_005245885.1 |
XM_005245830.3 |
1794 |
Missense Mutation |
AGG,GGG |
R513G |
XP_005245887.1 |
XM_005245832.3 |
1794 |
Missense Mutation |
AGG,GGG |
R513G |
XP_005245889.1 |
XM_006710555.3 |
1794 |
Missense Mutation |
AGG,GGG |
R522G |
XP_006710618.1 |
XM_011541198.2 |
1794 |
Missense Mutation |
AGG,GGG |
R513G |
XP_011539500.1 |
XM_017000954.1 |
1794 |
Missense Mutation |
AGG,GGG |
R522G |
XP_016856443.1 |
XM_017000955.1 |
1794 |
Missense Mutation |
AGG,GGG |
R522G |
XP_016856444.1 |
XM_017000956.1 |
1794 |
Missense Mutation |
AGG,GGG |
R521G |
XP_016856445.1 |
XM_017000957.1 |
1794 |
Missense Mutation |
AGG,GGG |
R502G |
XP_016856446.1 |
XM_017000958.1 |
1794 |
UTR 3 |
|
|
XP_016856447.1 |
XM_017000959.1 |
1794 |
UTR 3 |
|
|
XP_016856448.1 |
View Full Product Details