Product Details

SNP ID
rs149842844
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:220151641 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCAAGTACACAGTGTGGGTGGAA[G/T]TCTGGCAAGCAGCTCCATTCCTTCT
Phenotype
MIM: 612801 MIM: 609275
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IARS2 PubMed Links

Gene Details

Gene
IARS2
Gene Name
isoleucyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.

Gene
RAB3GAP2
Gene Name
RAB3 GTPase activating non-catalytic protein subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012414.3 4155 Missense Mutation ATT,CTT I1331L NP_036546.2

View Full Product Details