Product Details

SNP ID
rs150120731
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:111414789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGAGGGGACAGTCACCTTTTCA[C/T]GGGCCACAGCCTTCCTTCTAGGGGC
Phenotype
MIM: 603578
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OVGP1 PubMed Links

Gene Details

Gene
OVGP1
Gene Name
oviductal glycoprotein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002557.3 1768 Missense Mutation CAT,CGT H571R NP_002548.3

View Full Product Details