Product Details

SNP ID
rs150868852
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:84479496 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTAATGTTCACGCGGTGGAAAC[A/G]GCAGCAGCGGAAGGTACGCGAGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPF1 PubMed Links

Gene Details

Gene
RPF1
Gene Name
ribosome production factor 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025065.6 260 Missense Mutation CAG,CGG Q72R NP_079341.2

View Full Product Details