Product Details

SNP ID
rs141290046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32360171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTACAGATTTATAAAAACTGAAG[A/G]AATTTGGGCTACAGAATTTTCATTG
Phenotype
MIM: 612990
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ASXL1 PubMed Links

Gene Details

Gene
ASXL1
Gene Name
additional sex combs like 1, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164603.1 919 Intron NP_001158075.1
NM_015338.5 919 Intron NP_056153.2
XM_006723727.3 919 Intron XP_006723790.1
XM_006723728.3 919 Intron XP_006723791.1
XM_006723730.3 919 Intron XP_006723793.1
XM_006723732.2 919 Intron XP_006723795.1
XM_006723733.1 919 Intron XP_006723796.1
XM_011528648.2 919 Intron XP_011526950.1
XM_011528652.2 919 UTR 5 XP_011526954.1
XM_017027704.1 919 UTR 5 XP_016883193.1
XM_017027705.1 919 Intron XP_016883194.1
XM_017027706.1 919 Intron XP_016883195.1

View Full Product Details