Product Details

SNP ID
rs142050922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:23354116 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGAGTTTGTCAATGTCTACTA[C/T]ACCACCATGGATAAGCGGCGGCGTT
Phenotype
MIM: 613842 MIM: 605811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GZF1 PubMed Links

Gene Details

Gene
GZF1
Gene Name
GDNF inducible zinc finger protein 1
There are no transcripts associated with this gene.

Gene
LINC01431
Gene Name
long intergenic non-protein coding RNA 1431
There are no transcripts associated with this gene.

Gene
NXT1
Gene Name
nuclear transport factor 2 like export factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013248.2 231 Silent Mutation TAC,TAT Y25Y NP_037380.1
XM_011529230.2 231 Silent Mutation TAC,TAT Y25Y XP_011527532.1

View Full Product Details